Anti-BRCA1 Mouse Monoclonal Antibody [clone: GLK-2]

Supplier: Diagnostic Biosystems

MOB424 MOB424-05
99990-784EA 1402.04 USD
99990-784 95024-252
Anti-BRCA1 Mouse Monoclonal Antibody [clone: GLK-2]
Antibodies
BRCA1 recognizes a 22kDa protein. This antibody stains 94% of ovarian carcinomas with a mutation in exon 11. However, staining was absent in 100% of tumors with mutations other than exon 11.

Type:
Antigen: BRCA1
Clonality: Monoclonal
Clone: GLK-2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR