Anti-ACT1 Mouse Monoclonal Antibody [clone: 1A4]

Supplier: Diagnostic Biosystems

MOB001-05 PDM003 MOB001
95024-212EA 381.06 USD
95024-212 99990-820 99990-164
Anti-ACT1 Mouse Monoclonal Antibody [clone: 1A4]
Antibodies
This antibody is specific to α-smooth muscle isoform of actin. It reacts with smooth muscle cells of vessels and different parenchymes. This antibody does not cross-react with β and γ-cytoplasmic, α-sarcomeric and α-myocardial actin isoforms.

Type:
Antigen: ACT1
Clonality: Monoclonal
Clone: 1A4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR