Anti-AFP Mouse Monoclonal Antibody [clone: C3]

Supplier: Diagnostic Biosystems

MOB129 PDM057 MOB129-05
99990-330EA 415.9 USD
99990-330 99990-904 95024-226
Anti-AFP Mouse Monoclonal Antibody [clone: C3]
Antibodies
α-fetoprotein (AFP) is a single chain glycoprotein of 70 kDa that is normally expressed in the fetal liver, gastrointestinal tract and yolk sac. This antibody recognizes an epitope of α-fetoprotein present in human and several other species.

Type:
Antigen: AFP
Clonality: Monoclonal
Clone: C3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR