Anti-alpha-Actinin Mouse Monoclonal Antibody [clone: EA-53]

Supplier: Diagnostic Biosystems

MOB227 MOB227-05
99990-450EA 601.32 USD
99990-450 95024-224
Anti-alpha-Actinin Mouse Monoclonal Antibody [clone: EA-53]
Antibodies
This antibody is specific for α-skeletal muscle actinin and α-cardiac actinin. It stains Z lines and dots in stress fibers of myotubes in skeletal and cardiac muscle but not in non-sarcomeric muscle elements.

Type:
Antigen: alpha-Actinin
Clonality: Monoclonal
Clone: EA-53
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR