Anti-A1AT Rabbit Polyclonal Antibody

Supplier: Diagnostic Biosystems

RP048-05 PDR021 RP048
95024-902EA 184.11 USD
95024-902 99991-082 99991-284
Anti-A1AT Rabbit Polyclonal Antibody
Antibodies
The antibody is specific against human α-1-antitrypsin. This antibody stains macrophages in human tissues and does not stain any other cell type.

Type:
Antigen: A1AT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR