N-tert-Butyl-6-chloronicotinamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: N-(tert-Butyl)-6-chloronicotinamide

M-Clarity™ quality level = MQ200

8.43921.0005 8.43921.0025
EM8.43921.0005CS 51126 USD
EM8.43921.0005 EM8.43921.0025
N-tert-Butyl-6-chloronicotinamide for synthesis, Sigma-Aldrich®
N-tert-Butyl-6-chloronicotinamide
Formula: C₁₀H₁₃ClN₂O
MW: 212.677 g/mol
MDL Number: MFCD08460992
CAS Number: 115309-58-5

Order Now

Specification Test Results

Melting Point 108 - 110 °C
Molar Mass 212.68 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR