3,6-Dichloropyridazine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.42712.0025 8.42712.0100
EM8.42712.0025CS 8703 USD
EM8.42712.0025 EM8.42712.0100
3,6-Dichloropyridazine for synthesis, Sigma-Aldrich®
3,6-Dichloropyridazine
Formula: C₄H₂Cl₂N₂
Boiling Pt: 89…91 °C (0.2 mmHg)
Melting Pt: 70 °C
Density: 1.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006466
CAS Number: 141-30-0
EINECS: 205-478-6
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 0.0012 g/l (20 °C)
Melting Point 72 °C
Molar Mass 148.98 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR