(3-Chloropropyl)trichlorosilane ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Trichloro(3-chloropropyl)silane
M-Clarity™ quality level = MQ200
8.42016.0005
8.42016.0025
EM8.42016.0005CS
56716
USD
EM8.42016.0005
EM8.42016.0025
(3-Chloropropyl)trichlorosilane ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
(3-Chloropropyl)trichlorosilane
Formula:
ClH₂CCH₂CH₂SiCl₃ MW: 211.98 g/mol Boiling Pt: 176…177 °C (1013 hPa) Density: 1.359 g/cm³ (20 °C) |
MDL Number:
MFCD00013694 CAS Number: 2550-06-3 EINECS: 219-844-8 UN: 2987 ADR: 8,II |
Specification Test Results
Assay (GC, area%) | ≥ 98.0 % (a/a) |
Density (d 20 °C/ 4 °C) | 1.358 - 1.362 |
Identity (IR) | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...