4-Amino-1-naphthalenesulfonic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 4-Aminonaphthalene-1-sulphonic acid, 4-Amino-α-naphthalenesulphonic acid

M-Clarity™ quality level = MQ200

8.41535.0050 8.41535.0100
EM8.41535.0050CS 34138 USD
EM8.41535.0050 EM8.41535.0100
4-Amino-1-naphthalenesulfonic acid for synthesis, Sigma-Aldrich®
4-Amino-1-naphthalenesulphonic acid
Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
Boiling Pt: 434 °C (1013 hPa)
Density: 1.67 g/cm³ (20 °C)
MDL Number: MFCD00004027
CAS Number: 84-86-6
EINECS: 201-567-9
UN: 2585
ADR: 8,III
Merck Index: 12,06490

Order Now

Specification Test Results

Solubility 0.31 g/l (20 °C)
Melting Point >300 °C
Molar Mass 223.24 g/mol
Density 1.67 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR