4'-Chloroacetanilide for synthesis, Sigma-Aldrich®

Supplier: MERCK, S.A. DE C.V. SV

Synonyms: 4-Chloroacetanilide, Paracetamol impurity J, N-(4-Chlorophenyl)acetamide

M-Clarity™ quality level = MQ200

8.41480.0050 8.41480.0250
EM8.41480.0050CS 839 USD
EM8.41480.0050 EM8.41480.0250
4'-Chloroacetanilide for synthesis, Sigma-Aldrich®
4'-Chloroacetanilide
Formula: C₈H₈ClNO
MW: 169.61 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 179…180 °C
Density: 1.385 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000612
CAS Number: 539-03-7
EINECS: 208-707-8
Merck Index: 13,02128

Order Now

Specification Test Results

Solubility 0.95 g/l (65 °C) (experimental)
Melting Point 179 - 180 °C
Molar Mass 169.61 g/mol
Boiling Point 333 °C (1013 hPa)
Density 1.385 g/cm3 (22 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR