N,N-Dimethylethylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: DMEA

M-Clarity™ quality level = MQ200

8.40018.0100 8.40018.0500
EM8.40018.0100CS 492 USD
EM8.40018.0100 EM8.40018.0500
N,N-Dimethylethylamine for synthesis, Sigma-Aldrich®
N,N-Dimethylethylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 36 °C (1013 hPa)
Melting Pt: –140 °C
Density: 0.675 g/cm³ (20 °C)
Flash Pt: –45 °C
MDL Number: MFCD00009039
CAS Number: 598-56-1
EINECS: 209-940-8
UN: 2733
ADR: 3 (8),II

Order Now

Specification Test Results

Solubility (20 °C) miscible
Melting Point -140 °C
Molar Mass 73.14 g/mol
Boiling Point 36.5 °C (1013 hPa)
Vapor Pressure 527 hPa (20 °C)
Flash Point -45.5 °C
Explosion Limit 0.9 - 11.2 %(V)
Density 0.675 g/cm3 (20 °C)
pH 12.0 (50 g/l, H2O, 20 °C)
Ignition Point 190 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR