Carbazole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.20255.0005 8.20255.0250
EM8.20255.0005CS 30984 USD
EM8.20255.0005 EM8.20255.0250
Carbazole for synthesis, Sigma-Aldrich®
Carbazole
Formula: C₁₂H₉N
MW: 167.21 g/mol
MDL Number: MFCD00004960
CAS Number: 86-74-8
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility 0.1 g/l (20 °C)
Melting Point 244 - 246 °C
Molar Mass 167.2 g/mol
Bulk Density 620 kg/m3
Boiling Point 355 °C (1013 hPa)
Flash Point 220 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR