Xenon difluoride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18888.0001 8.18888.0005
EM8.18888.0001CS 1881.81 USD
EM8.18888.0001 EM8.18888.0005
Xenon difluoride for synthesis, Sigma-Aldrich®
Xenon difluoride
Formula: F₂Xe
MDL Number: MFCD00040538
CAS Number: 13709-36-9
UN: 3085
ADR: 5.1 (8),II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 130 - 135 °C (sublimed)
Molar Mass 169.29 g/mol
Vapor Pressure 3.8 hPa (25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR