Methyl nicotinate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
3-Picolinic acid methyl ester, 3-(Methoxycarbonyl)pyridine, Pyridine-3-carboxylic acid methyl ester, Nicotinic acid methyl ester, 3-Pyridinecarboxylic acid methyl ester, Methyl 3-pyridinecarboxylate
M-Clarity™ quality level = MQ200
8.18713.0100
8.18713.0500
EM8.18713.0100CS
1175
USD
EM8.18713.0100
EM8.18713.0500
Methyl nicotinate for synthesis, Sigma-Aldrich®
Methyl nicotinate
Formula:
C₇H₇NO₂ MW: 137.14 g/mol |
MDL Number:
MFCD00006388 CAS Number: 93-60-7 |
Specification Test Results
Solubility | 48 g/l (20 °C) |
Melting Point | 39 °C |
Molar Mass | 137.14 g/mol |
Bulk Density | 600 kg/m3 |
Boiling Point | 209 °C (1013 hPa) |
Vapor Pressure | 0.4 hPa (25 °C) |
Flash Point | 95 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...