N-Alkyl-N,N-dimethyl-N-benzylammonium chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Benzyldimethylmyristylammonium chloride

M-Clarity™ quality level = MQ200

8.14363.0100 8.14363.0500
EM8.14363.0100CS 2902 USD
EM8.14363.0100 EM8.14363.0500
N-Alkyl-N,N-dimethyl-N-benzylammonium chloride for synthesis, Sigma-Aldrich®
N-Alkyl-N,N-dimethyl-N-benzylammonium chloride
Formula: (C₆H₅CH₂)(CH₃)₂N(C₁₂⁻C₁₄Alkyl)⁺Cl⁻
Melting Pt: 73 °C
Storage Temperature: Ambient
CAS Number: 85409-22-9
EINECS: 287-089-1
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Assay (argentometric) ≥ 98.0 %
Melting range (upper value) ≤ 73 °C
Identity (IR) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR