Tetrabutyltin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.08278.0025 8.08278.0100
EM8.08278.0025CS 2275.73 USD
EM8.08278.0025 EM8.08278.0100
Tetrabutyltin for synthesis, Sigma-Aldrich®
Tetrabutyltin
Formula: C₁₆H₃₆Sn
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –97 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009422
CAS Number: 1461-25-2
EINECS: 215-960-8
UN: 2788
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 0.008 g/l (20 °C)
Melting Point -97 °C
Molar Mass 347.15 g/mol
Boiling Point 140 °C (10 hPa)
Vapor Pressure 0.0014 hPa (25 °C)
Flash Point >100 °C
Refractive Index 1.4742 (20 °C, 589 nm)
Density 1.05 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR