Malononitrile for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.06189.0100 8.06189.0250
EM8.06189.0100CS 877 USD
EM8.06189.0100 EM8.06189.0250
Malononitrile for synthesis, Sigma-Aldrich®
Malononitrile
Formula: C₃H₂N₂
MW: 66.06 g/mol
MDL Number: MFCD00001883
CAS Number: 109-77-3
UN: 2647
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 133 g/l (20 °C)
Melting Point 32 - 34 °C
Molar Mass 66.06 g/mol
Boiling Point 220 °C (1013 hPa)
Vapor Pressure 1 hPa (50 °C)
Flash Point 112 °C
Density 1.05 g/cm3 (20 °C)
Ignition Point 365 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR