Potassium cyanate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Cyanic acid potassium salt

M-Clarity™ quality level = MQ200

8.04957.0100 8.04957.1000
EM8.04957.0100CS 7551 USD
EM8.04957.0100 EM8.04957.1000
Potassium cyanate for synthesis, Sigma-Aldrich®
Potassium cyanate
Formula: CKNO
MW: 81.12 g/mol
MDL Number: MFCD00011411
CAS Number: 590-28-3

Order Now

Specification Test Results

Solubility 750 g/l (20 °C)
Melting Point 314 - 316 °C
Molar Mass 81.12 g/mol
Bulk Density 870 kg/m3
Density 2.056 g/cm3 (20 °C)
pH 10 (50 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR