4-Bromobenzaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.04146.0010 8.04146.0100
EM8.04146.0010CS 5306 USD
EM8.04146.0010 EM8.04146.0100
4-Bromobenzaldehyde for synthesis, Sigma-Aldrich®
4-Bromobenzaldehyde
Formula: C₇H₅BrO
MDL Number: MFCD00003377
CAS Number: 1122-91-4

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 55 - 58 °C
Molar Mass 185.02 g/mol
Bulk Density 670 kg/m3
Boiling Point 123 - 125 °C (31 hPa)
Flash Point 108 °C
Density 1.85 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR