Vinyl acetate stabilized for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Acetic acid vinyl ester
M-Clarity™ quality level = MQ200
8.03184.0100
8.03184.1000
EM8.03184.0100CS
524
USD
EM8.03184.0100
EM8.03184.1000
Vinyl acetate stabilized for synthesis, Sigma-Aldrich®
Vinyl acetate
Formula:
C₄H₆O₂ MW: 86.09 g/mol Boiling Pt: 72.3 °C (1013 hPa) Melting Pt: -100 °C Density: 0.932 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008713 CAS Number: 108-05-4 EINECS: 203-545-4 UN: 1301 ADR: 3,II Merck Index: 12,10130 |
Specification Test Results
Solubility | 20 g/l (20 °C) |
Melting Point | -93 °C |
Molar Mass | 86.09 g/mol |
Boiling Point | 71 - 73 °C (1013 hPa) |
Vapor Pressure | 120 hPa (20 °C) |
Flash Point | -8 °C |
Refractive Index | 1.3954 (20 °C, 589 nm) |
Explosion Limit | 2.6 - 13.4 %(V) |
Density | 0.93 g/cm3 (20 °C) |
pH | 7 (20 g/l, H2O, 20 °C) |
Ignition Point | 385 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...