4-Chloro-m-cresol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 4-Chloro-3-methylphenol, 2-Chloro-5-hydroxytoluene, 1-Chloro-4-hydroxy-2-methylbenzene, p-Chloro m-cresol

M-Clarity™ quality level = MQ200

8.02295.0005 8.02295.0250 8.02295.1000 8.02295.9050
EM8.02295.0005CS 20010.4 USD
EM8.02295.0005 EM8.02295.0250 EM8.02295.1000 EM8.02295.9050
4-Chloro-m-cresol ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
4-Chloro-m-cresol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 235…239 °C (1013 hPa)
Melting Pt: 63…65 °C
Density: 0.9 g/cm³ (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00002323
CAS Number: 59-50-7
EINECS: 200-431-6
UN: 3437
ADR: 6.1,II
Merck Index: 13,02152

Order Now

Specification Test Results

Solubility 3.9 g/l (20 °C)
Melting Point 63 - 65 °C
Molar Mass 142.58 g/mol
Bulk Density 800 kg/m3
Boiling Point 235 - 239 °C (1013 hPa)
Vapor Pressure < - 0.1 hPa (20 °C)
Flash Point 118 °C
Density 1.37 g/cm3 (20 °C)
pH 6.5 (1 g/l, H2O, 20 °C)
Ignition Point 590 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR