2-Bromo-2-methylpropane stabilised for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
tert-Butyl bromide
Stabilization:
Stabilised with potassium carbonate
M-Clarity™ quality level = MQ200
8.01966.0250
8.01966.1000
EM8.01966.0250CS
1035
USD
EM8.01966.0250
EM8.01966.1000
2-Bromo-2-methylpropane stabilised for synthesis, Sigma-Aldrich®
2-Bromo-2-methylpropane
Formula:
C₄H₉Br Boiling Pt: 73 °C (1013 hPa) Melting Pt: -20 °C Density: 1.2125 g/cm³ (20 °C) Flash Pt: 16 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000125 CAS Number: 507-19-7 EINECS: 208-065-9 UN: 2342 ADR: 3,II Merck Index: 13,01554 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | -20 °C |
Molar Mass | 137.01 g/mol |
Boiling Point | 72 - 74 °C |
Vapor Pressure | 179.9 hPa (25 °C) |
Flash Point | 16 °C |
Refractive Index | 1.4279 (20 °C, 589 nm) |
Density | 1.22 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...