Phthalonitrile ≥98% (by GC) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
1,2-Benzenedicarbonitrile, Benzene-1,2-dicarbonitrile, 1,2-Dicyanobenzene
M-Clarity™ quality level = MQ200
8.00593.0100
8.00593.0500
8.00593.9050
EM8.00593.0100CS
520.79
USD
EM8.00593.0100
EM8.00593.0500
EM8.00593.9050
Phthalonitrile ≥98% (by GC) for synthesis, Sigma-Aldrich®
Phthalonitrile
Formula:
C₈H₄N₂ MW: 128.13 g/mol Boiling Pt: 305 °C (1013 hPa) Melting Pt: 138…141 °C Density: 1.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001771 CAS Number: 91-15-6 EINECS: 202-044-8 UN: 3439 ADR: 6.1,III |
Specification Test Results
Solubility | 0.56 g/l (25 °C) |
Melting Point | 138 - 141 °C |
Molar Mass | 128.13 g/mol |
Bulk Density | 500 kg/m3 |
Vapor Pressure | 0.04 hPa (20 °C) |
Density | 1.24 g/cm3 (20 °C) |
pH | 7 (H2O) |
Ignition Point | >580 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...