Germanium(IV) oxide ≥99%, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Germanium dioxide

M-Clarity™ quality level = MQ200

1.12177.0005 1.12177.0025
EM1.12177.0005CS 6500 USD
EM1.12177.0005 EM1.12177.0025
Germanium(IV) oxide ≥99%, Sigma-Aldrich®
Germanium(IV) oxide
Formula: GeO₂
MW: 104.61 g/mol
MDL Number: MFCD00011030
CAS Number: 1310-53-8

Order Now

Specification Test Results

Solubility 4.5 g/l (25 °C)
Melting Point 1115 °C
Molar Mass 104.61 g/mol
Bulk Density 1430 kg/m3
Density 4.23 g/cm3 (25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR