Atorvastatin ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: 2-​(4-​fluorophenyl)-​βR,​δR-​dihydroxy-​5-​(1-​methylethyl)-​3-​phenyl- ​4-​[(phenylamino)carbonyl]-​1H-​pyrrole-​1-​heptanoic acid, (3R,5R)-7-(2-(4-Fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoic acid

075942-1G 075942-5G
102543-738EA 146.15 USD
102543-738 102543-740
Atorvastatin ≥95%
Atorvastatin
Formula: C₃₃H₃₅FN₂O₅
MDL Number: MFCD00899261
CAS Number: 134523-00-5

Order Now

Specification Test Results

Molecular Weight 558.66


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR