Clorox® Healthcare® Bleach Germicidal Wipes
Supplier: Janitorial Supplies
CLO30358CT CLO35309CT CLO30359CT
89492-610CS 265.56 USD
89492-610 89492-612 89492-614
Clorox® Healthcare® Bleach Germicidal Wipes
Wipes Disposable Wipes and Towels Presaturated Wipes
Healthcare professionals trust Clorox® Healthcare® bleach germicidal wipes to help them overcome infection control challenges in a constantly changing healthcare environment.
  • No pre-cleaning required
  • EPA registered to kill a total of 58 microorganisms
  • Proven efficacy against outbreak-causing viruses Influenza A and B viruses, Measles virus and Norovirus
  • Kill Healthcare associated infections-causing pathogens including Clostridioides difficile spores, MRSA and Vancomycin-resistant enterococci in three minutes
  • Broad Spectrum disinfection efficacy against over 58 pathogens
  • Three minute contact time to kill Clostridioides difficile spores
  • Meets CDC and OSHA recommendations
  • Feature an anticorrosion agent to ensure compatibility with healthcare surfaces
  • Tested for surface safety on common healthcare and office surfaces and equipment
Clorox Healthcare Bleach Germicidal clinical wipes are ready to use, one step cleaner containing a 1:10 bleach dilution to meet CDC and OSHA guidelines for disinfecting surfaces in healthcare settings. Designed for daily disinfection, high-touch surfaces and medical equipment. These wipes kill 99.9999% of Clostridioides difficile spores.
These wipes are engineered minimize residue and leave less visible residue on surfaces vs. competitor bleach wipes. They are formulated with an anticorrosion agent and are compatible with common healthcare surfaces including stainless steel, glass, plastic, porcelain, glazed ceramic, laminated surfaces and vinyl. Intended for use in healthcare settings. Cleans bacteria, dust, fungus, germs, mildew, mold, soil, viruses. Unscented. For use in hospitals, nursing homes, and medical facilities. For use on acrylic, ceramic, chrome, fiberglass, glass, metal, plastic, tile, vinyl, and wood.
Certificaciones: EPA recommended testing standard (June 2014).