1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, Powder USP

Supplier: AVANTI POLAR LIPIDS INC MS

850457P25MG 850457P1G 850457P200MG 850457P500MG
100125-180EA 145 USD
100125-180 100126-580 100121-410 100127-972
1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, Powder USP
1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine
Formula: C₄₂H₈₂NO₈P
MW: 760.091 g/mol
MDL Number: MFCD00077387
CAS Number: 26853-31-6

Order Now

Specification Test Results

Molecular Weight 760.076
Exact Mass 759.578

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR