Copper(II) gluconate USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Copper(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic Acid Copper(II) Salt, Copper(II) D-gluconate, Copper Gluconate
C1334-2.5KG
C1334-125GM
C1334-25KGBL
C1334-500GM
95032-686EA
1238.75
USD
95032-686
95032-684
89078-772
95032-688
Copper(II) gluconate USP
Copper(II) gluconate
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₁₂H₂₂CuO₁₄ MW: 453.84 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00075297 CAS Number: 527-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...