4-Methyl-3-nitroanisole ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: 1-Methoxy-4-methyl-3-nitrobenzene, 4-Methoxy-2-nitrotoluene, Methyl 3-nitro-p-tolyl ether

074081-5G 074081-25G
102534-546EA 90.77 USD
102534-546 102833-670
4-Methyl-3-nitroanisole ≥95%
4-Methyl-3-nitroanisole
Formula: C₈H₉NO₃
Boiling Pt: 266…267°C (760mm)
Melting Pt: 17°C
Density: 1.207 g/cm3
MDL Number: MFCD00007173
CAS Number: 17484-36-5
UN: 2446
ADR: 6.1,III

Order Now

Specification Test Results

Boiling Point 266 to 267°
Melting Point 17°
Density 1.207


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR