2-Chlorobenzaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20276.0250 8.20276.1000
EM8.20276.0250CS 494 USD
EM8.20276.0250 EM8.20276.1000
2-Chlorobenzaldehyde for synthesis, Sigma-Aldrich®
2-Chlorobenzaldehyde
Formula: C₇H₅ClO
MW: 140.57 g/mol
MDL Number: MFCD00003304
CAS Number: 89-98-5
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Solubility 1.12 g/l (20 °C)
Melting Point 10 - 11.5 °C
Molar Mass 140.56 g/mol
Boiling Point 208 °C
Vapor Pressure 1.7 hPa (50 °C)
Flash Point 87 °C
Refractive Index 1.5658 (20 °C, 589 nm)
Density 1.25 g/cm3 (20 °C)
pH 2.9 (H2O) (saturated aqueous solution)
Ignition Point 397 °C
Hazard Class 8
Hazard Class Description Corrosive material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR