Ethylamine ≥70% in aqueous solution for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Monoethylamine, Aminoethane, Ethanamine
M-Clarity™ quality level = MQ200
8.20024.1000
8.20024.2500
EM8.20024.1000CS
354
USD
EM8.20024.1000
EM8.20024.2500
Ethylamine ≥70% in aqueous solution for synthesis, Sigma-Aldrich®
Ethylamine
Formula:
C₂H₇N |
MDL Number:
MFCD00008160 CAS Number: 75-04-7 UN: 2270 ADR: 3 (8),II |
Specification Test Results
Solubility | (20 °C) soluble |
Boiling Point | 39 °C (1013 hPa) |
Vapor Pressure | 460 hPa (20 °C) |
Flash Point | -26 °C |
Explosion Limit | 3.0 - 12.8 %(V) |
Density | 0.81 g/cm3 (20 °C) |
pH | >14 (H2O, 20 °C) |
Ignition Point | 375 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...