N-Chlorosuccinimide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1-Chloropyrrolidine-2,5-dione

M-Clarity™ quality level = MQ200

8.02811.0005 8.02811.0250
EM8.02811.0005CS 40830 USD
EM8.02811.0005 EM8.02811.0250
N-Chlorosuccinimide for synthesis, Sigma-Aldrich®
N-Chlorosuccinimide
Formula: C₄H₄ClNO₂
MW: 133.53 g/mol
Boiling Pt: 216.5 °C (1013 hPa)
Melting Pt: 148 - 151 °C
Density: 1.65 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005511
CAS Number: 128-09-6
EINECS: 204-878-8
UN: 3261
ADR: 8,II
Merck Index: 13,02183

Order Now

Specification Test Results

Solubility 14 g/l (20 °C)
Melting Point 146 - 151 °C
Molar Mass 133.53 g/mol
Bulk Density 600 kg/m3
Boiling Point 216.5 °C
Flash Point >110 °C
Density 1.65 g/cm3 (20 °C)
pH (20 g/l, H2O) acidic
Hazard Class 8
Hazard Class Description Corrosive material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR