2-Imidazolidinone hemihydrate ≥97.0% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2-Imidazolidone hemihydrate

M-Clarity™ quality level = MQ200

8.04789.0100 8.04789.0500
EM8.04789.0100CS 5315.71 USD
EM8.04789.0100 EM8.04789.0500
2-Imidazolidinone hemihydrate ≥97.0% for synthesis, Sigma-Aldrich®
2-Imidazolidinone hemihydrate
Formula: C₃H₆N₂O·0.5H₂O
MW: 95.10 (86.09 anhy)
MDL Number: MFCD02662355
CAS Number: 121325-67-5
EINECS: 204-436-4

Order Now

Specification Test Results

Solubility 950 g/l (20 °C)
Melting Point 58 °C
Molar Mass 95.10 g/mol
Bulk Density 720 kg/m3
Boiling Point 132.6 °C (1013 hPa) (anhydrous substance)
Vapor Pressure < - 24 hPa (20 °C)
Flash Point 265 °C
Density 1.3 g/cm3 (20 °C)
pH 9 - 11 (100 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR