Methyl hydroxyethylcellulose for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Tylose, Methyl 2-hydroxyethyl cellulose

M-Clarity™ quality level = MQ200

8.22069.0100 8.22069.0500
EM8.22069.0100CS 3329 USD
EM8.22069.0100 EM8.22069.0500
Methyl hydroxyethylcellulose for synthesis, Sigma-Aldrich®
Methyl hydroxyethylcellulose
MDL Number: MFCD00147597
CAS Number: 9032-42-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Bulk Density 350 kg/m3
Density 1.28 - 1.30 g/cm3 (20 °C)
pH (10 g/l, H2O, 20 °C) neutral
Ignition Point >360 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR