Pentyl nitrite (mixed isomers ) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Amyl Nitrite, Pentyl Nitrite

M-Clarity™ quality level = MQ200

8.20966.0005 8.20966.0250
EM8.20966.0005CS 14633 USD
EM8.20966.0005 EM8.20966.0250
Pentyl nitrite (mixed isomers ) for synthesis, Sigma-Aldrich®
Pentyl nitrite (mixed isomers )
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
MDL Number: MFCD00036132
CAS Number: 463-04-7
UN: 1113
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Molar Mass 117.14 g/mol
Boiling Point 104 °C (1013 hPa)
Vapor Pressure 65 hPa (20 °C)
Flash Point -40 °C
Density 0.86 g/cm3 (20 °C)
Ignition Point 205 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR