Triethylenetetramine mixture of isomers ≥95% for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
N,N'-Bis(2-aminoethyl)-1,2-ethanediamine, Trientine, DEH 24, EPH 925
M-Clarity™ quality level = MQ200
8.14392.0100
8.14392.0500
EM8.14392.0100CS
570
USD
EM8.14392.0100
EM8.14392.0500
Triethylenetetramine mixture of isomers ≥95% for synthesis, Sigma-Aldrich®
Triethylenetetramine
Formula:
C₆H₁₈N₄ MW: 146.24 g/mol |
MDL Number:
MFCD00008169 CAS Number: 112-24-3 UN: 2259 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 12 °C |
Molar Mass | 146.23 g/mol |
Boiling Point | 270 - 300 °C (1013 hPa) |
Vapor Pressure | < - 0.01 hPa (20 °C) |
Flash Point | 129 °C |
Refractive Index | 1.4971 (20 °C, 589 nm) |
Explosion Limit | 0.7 - 7.2 %(V) |
Density | 0.98 g/cm3 (20 °C) |
pH | 10 - 11 (10 g/l, H2O, 20 °C) |
Ignition Point | 335 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...