N,N,N-Trimethylanilinium chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Trimethylphenylammonium chloride, Phenyltrimethylammonium chloride

M-Clarity™ quality level = MQ200

8.08428.0100 8.08428.0500
EM8.08428.0100CS 9888 USD
EM8.08428.0100 EM8.08428.0500
N,N,N-Trimethylanilinium chloride for synthesis, Sigma-Aldrich®
N,N,N-Trimethylanilinium chloride
Formula: C₉H₁₄ClN
MW: 171.67 g/mol
MDL Number: MFCD00011790
CAS Number: 138-24-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 333 g/l (20 °C)
Melting Point 246 - 247 °C
Molar Mass 171.66 g/mol
Bulk Density 540 kg/m3
Explosion Limit 1 - 6 %(V)
pH 4.5 (333 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR