3-Mercaptopropionic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.05967.0100 8.05967.0250
EM8.05967.0100CS 741 USD
EM8.05967.0100 EM8.05967.0250
3-Mercaptopropionic acid for synthesis, Sigma-Aldrich®
3-Mercaptopropionic acid
Formula: C₃H₆O₂S
MW: 106.15 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 17…19 °C
Density: 1.2199 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00004897
CAS Number: 107-96-0
EINECS: 203-537-0
UN: 2922
ADR: 8 (6.1),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 17 - 19 °C
Molar Mass 106.14 g/mol
Boiling Point 110 °C (20 hPa)
Vapor Pressure 0.053 hPa (20 °C)
Flash Point 93 °C
Refractive Index 1.4911 (20 °C, 589 nm)
Explosion Limit 1.60 %(V)
Density 1.22 g/cm3 (20 °C)
pH 2 (120 g/l, H2O, 20 °C)
Ignition Point 350 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR