3-Methylbutanone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.05966.0250 8.05966.1000
EM8.05966.0250CS 744 USD
EM8.05966.0250 EM8.05966.1000
3-Methylbutanone for synthesis, Sigma-Aldrich®
3-Methylbutanone
Formula: C₅H₁₀O
Boiling Pt: 94 °C (1013 hPa)
Melting Pt: –92 °C
Density: 0.8046 g/cm³ (20 °C)
Flash Pt: –3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008919
CAS Number: 563-80-4
EINECS: 209-264-3
UN: 2397
ADR: 3,II
Merck Index: 12,06164

Order Now

Specification Test Results

Solubility 6 g/l (20 °C)
Melting Point -92 °C
Molar Mass 86.13 g/mol
Boiling Point 93 - 95 °C (1013 hPa)
Vapor Pressure 53.33 hPa (19.9 °C)
Flash Point -1 °C
Refractive Index 1.388 (20 °C, 589 nm)
Explosion Limit 1.5 - 9 %(V)
Density 0.80 g/cm3 (20 °C)
Ignition Point 475 °C
Evaporation number 5.6


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR