2-Chloroethyldimethylammonium chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N-(2-Chloroethyl)-N,N-Dimethylammonium Chloride, 2-Dimethylaminoethyl chloride hydrochloride

M-Clarity™ quality level = MQ200

8.02448.0005 8.02448.0250 8.02448.1000
EM8.02448.0005CS 30209 USD
EM8.02448.0005 EM8.02448.0250 EM8.02448.1000
2-Chloroethyldimethylammonium chloride for synthesis, Sigma-Aldrich®
2-Chloroethyldimethylammonium chloride
Formula: C₄H₁₁Cl₂N
MW: 144.04 g/mol
MDL Number: MFCD00012516
CAS Number: 4584-46-7
EINECS: 224-970-1
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 2000 g/l (20 °C)
Melting Point 205 - 208 °C
Molar Mass 144.04 g/mol
Ignition Point 220 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR