Mercury(II) sulfate, Extra Pure, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Mercury sulphate, Mercuric sulphate
M-Clarity™ quality level = MQ200
1.04481.0250
1.04481.1000
EM1.04481.0100CS
4147
USD
EM1.04481.0100
EM1.04481.0250
EM1.04481.1000
Mercury(II) sulfate, Extra Pure, Supelco®
Mercury(II) sulphate
Formula:
HgSO₄ MW: 296.65 g/mol Melting Pt: 850 °C Density: 6.47 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011047 CAS Number: 7783-35-9 EINECS: 231-992-5 UN: 1645 ADR: 6.1,II Merck Index: 13,05914 |
Specification Test Results
Identity | passes test |
Chloride (Cl) | ≤ 0.003 % |
Fe (Iron) | ≤ 0.005 % |
Mercury (I) (as Hg) | ≤ 0.1 % |
Substances not reducing by formic acid (residue on ignition) | ≤ 0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...