Nitric acid 65%, extra pure, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
1.00443.2500
1.00443.9025
EM1.00443.2500CS
414
USD
EM1.00443.2500
EM1.00443.9025
Nitric acid 65%, extra pure, Supelco®
Nitric acid
Formula:
HNO₃ Boiling Pt: 120.5 °C (1013 hPa) Melting Pt: -42 °C Density: 1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 EINECS: 231-714-2 UN: 2031 ADR: 8,II Merck Index: 13,06608 |
Specification Test Results
Appearance of solution | passes test |
As (Arsenic) | ≤ 0.0001 % |
Assay (alkalimetric) | 64.3 - 66.4 % |
Ca (Calcium) | ≤ 0.001 % |
Chloride (Cl) | ≤ 0.0003 % |
Date of expiry | see product label |
Evaporation residue | ≤ 0.01 |
Fe (Iron) | ≤ 0.0004 % |
Heavy metals (as Pb) | ≤ 0.0005 % |
Identity | passes test |
Iodate, Bromate | passes test |
NH₄ (Ammonium) | ≤ 0.001 % |
Nitrogen oxides (as N₂O₃) | ≤ 0.003 % |
Residual solvents (ICH Q3C) | ...excluded by production process |
Sulfate (SO₄) | ≤ 0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...