Barbituric acid, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

1.00132.0025 1.00132.0100
EM1.00132.0025CS 23536.53 USD
EM1.00132.0025 EM1.00132.0100
Barbituric acid, EMSURE® for analysis, Supelco®
Barbituric acid

1.00132.0100 replaces and is the same as BX0035-3. Part # Change only, Same Pack Size, Same Specifications.

Formula: C₄H₄N₂O₃
Boiling Pt: 441 °C (1013 hPa)
Melting Pt: 250…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00006666
CAS Number: 67-52-7
EINECS: 200-658-0
Merck Index: 13,00966

Order Now

Specification Test Results

Solubility 11.45 g/l (25 °C)
Melting Point 250 - 252 °C (decomposition)
Molar Mass 128.08 g/mol
Bulk Density 720 kg/m3
pH 2 - 3 (50 g/l, H2O, 60 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR