SP Bel-Art Vikem® Vinyl Aprons, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H24604-0001 H24603-0001
47750-410EA 119.78 USD
47750-410 70680-334
SP Bel-Art Vikem® Vinyl Aprons, Bel-Art Products, a part of SP
Disposable Garments
Made from 4 mil thick vinyl film, these aprons have a stitched cotton binding securing all edges.
  • Vinyl fabrication
  • Non-absorbent
  • Chemically resistant
  • Reinforced with metal grommets
Strong tie backs are double reinforced with metal grommets through three layers of material at the waist.
Non-absorbent and chemically resistant, these aprons give excellent protection to personnel in hospitals, schools, laboratories and industrial plants. Yearly contracts on larger quantities at special discount prices are available.