Dulbecco's Phosphate-Buffered Saline, KPL Inc.
Supplier: KPL
5460-0030
5460-0031
95059-260EA
212.32
USD
95059-260
95059-262
Dulbecco's Phosphate-Buffered Saline, KPL Inc.
Media for Microbiology
Dehydrated Media for Microbiology
Dulbecco's Phosphate-Buffered Saline is a balanced salt solution ideal for use in sample preparation and as a wash buffer in immunoassay applications
As a modified Dulbecco's buffer, it contains no magnesium or calcium salts.