1,2,4-Trimethylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pseudocumene

012411-A3 012411-36 012411-B4 012411-A9
AA12411-A3EA 212 USD
AA12411-A3 AA12411-36 AA12411-B4 AA12411-A9
1,2,4-Trimethylbenzene 98%
1,2,4-Trimethylbenzene

1,2,4-Trimethylbenzene, 98%

Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 169…171 °C
Melting Pt: –44 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 48 °C (118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008527
CAS Number: 95-63-6
EINECS: 202-436-9
UN: 3295
ADR: 3,III
Merck Index: 12,08100

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR