Poloxamer 188

Supplier: Thermo Scientific Chemicals

J66087-36 J66087-A1
AAJ66087-36EA 227.73 USD
AAJ66087-36 AAJ66087-A1
Poloxamer 188
Poloxamer 188 (Lutrol® F68, Pluronic® F-68, Synperonic® F68)

Poloxamer 188


CAS No.: 9003-11-6 Physical Form: Solid Appearance: White MDL No.: MFCD00082049

Formula: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
Storage Temperature: Ambient
MDL Number: MFCD00082049
CAS Number: 9003-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR