7-Fluorooxindole 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7-Fluoroindolin-2-one

H61511-03 H61511-06 H61511-MD
AAH61511-03EA 129 USD
AAH61511-03 AAH61511-06 AAH61511-MD
7-Fluorooxindole 95%
7-Fluorooxindole

7-Fluorooxindole, 95%

Formula: C₈H₆FNO
MW: 151.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179608
CAS Number: 71294-03-6
EINECS: 676-237-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR