3-Bromo-2-fluoro-5-nitropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H61500-03 H61500-06 H61500-14
AAH61500-03EA 62.7 USD
AAH61500-03 AAH61500-06 AAH61500-14
3-Bromo-2-fluoro-5-nitropyridine 97%
3-Bromo-2-fluoro-5-nitropyridine

3-Bromo-2-fluoro-5-nitropyridine, 97%


Purity: 97% CAS No.: 1868-58-2 Molecular Formula: C5H2BrFN2O2 Formula Weight: 220.98 MDL No.: MFCD08703668

Formula: C₅H₂BrFN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08703668
CAS Number: 1868-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR