1-Bromo-2-fluoro-4-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-3-fluoronitrobenzene

H61494-14 H61494-22
AAH61494-14EA 103 USD
AAH61494-14 AAH61494-22
1-Bromo-2-fluoro-4-nitrobenzene 98%
1-Bromo-2-fluoro-4-nitrobenzene

1-Bromo-2-fluoro-4-nitrobenzene, 98%

Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Melting Pt: 88…94 °C
Storage Temperature: Ambient
MDL Number: MFCD05865088
CAS Number: 185331-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR